Learn More
2-Cyanobenzoic acid, 94%
CAS: 3839-22-3 | C8H5NO2 | 147.133 g/mol
Supplier: Thermo Scientific Chemicals H5023703
Description
2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications
| 2-Cyanobenzoic acid | |
| 210°C to 212°C | |
| MFCD00045796 | |
| o-cyanobenzoic acid, cyanobenzoic acid, benzoic acid, 2-cyano, 2-carboxybenzonitrile, 2-carboxy-benzonitrile, 2-cyanobenzoicacid, 2-cyano-benzoic acid, pubchem12513, acmc-1bn2v | |
| C1=CC=C(C(=C1)C#N)C(=O)O | |
| 147.133 | |
| 147.13 |
| 3839-22-3 | |
| C8H5NO2 | |
| 1 g | |
| DTNSDCJFTHMDAK-UHFFFAOYSA-N | |
| 2-cyanobenzoic acid | |
| 138061 | |
| 96% |
Chemical Identifiers
| 3839-22-3 | |
| 147.133 | |
| DTNSDCJFTHMDAK-UHFFFAOYSA-N | |
| 138061 | |
| C1=CC=C(C(=C1)C#N)C(=O)O |
| C8H5NO2 | |
| MFCD00045796 | |
| o-cyanobenzoic acid, cyanobenzoic acid, benzoic acid, 2-cyano, 2-carboxybenzonitrile, 2-carboxy-benzonitrile, 2-cyanobenzoicacid, 2-cyano-benzoic acid, pubchem12513, acmc-1bn2v | |
| 2-cyanobenzoic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only