CAS RN 3839-22-3
IUPAC Name:
2-cyanobenzoate
Synonyms:
2-cyanobenzoate
Molecular Weight (g/mol):
146.13
Molecular Formula:
C8H4NO2
InChi Key:
DTNSDCJFTHMDAK-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=CC=C1C#N
1
–
1
of
1
results
1
Promotions Available
2-Cyanobenzoic acid, 94%
2-Cyanobenzoic acid, 94%
CAS: 3839-22-3 Molecular Formula: C8H5NO2 Molecular Weight (g/mol): 147.133 MDL Number: MFCD00045796 InChI Key: DTNSDCJFTHMDAK-UHFFFAOYSA-N Synonym: o-cyanobenzoic acid,cyanobenzoic acid,benzoic acid, 2-cyano,2-carboxybenzonitrile,2-carboxy-benzonitrile,2-cyanobenzoicacid,2-cyano-benzoic acid,pubchem12513,acmc-1bn2v PubChem CID: 138061 IUPAC Name: 2-cyanobenzoic acid SMILES: C1=CC=C(C(=C1)C#N)C(=O)O