missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-4'-methylbiphenyl 99.0+%, TCI America™

Catalog No. C144610G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C144610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C144610G Supplier TCI America Supplier No. C144610G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 114772-53-1
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00151805
InChI Key ZGQVZLSNEBEHFN-UHFFFAOYSA-N
Synonym 2-cyano-4'-methylbiphenyl, 4'-methyl-2-cyanobiphenyl, 4'-methylbiphenyl-2-carbonitrile, 2'-cyano-4-methylbiphenyl, 2-p-tolyl benzonitrile, 2-4-methylphenyl benzonitrile, 2-cyano-4'-methyl biphenyl, 4'-methyl-1,1'-biphenyl-2-carbonitrile, unii-o417ti15ta, 4'-methyl-2-cyano diphenyl
PubChem CID 145512
IUPAC Name 4'-methyl-[1,1'-biphenyl]-2-carbonitrile
SMILES CC1=CC=C(C=C1)C1=CC=CC=C1C#N

Specifications

Melting Point 53°C
Quantity 10 g
Formula Weight 193.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-4′-methylbiphenyl
EINECSNumber (4)-1772
TSCA No