accessibility menu, dialog, popup

UserName

CH3N

CAS RN 114772-53-1

IUPAC Name: 4'-methyl-[1,1'-biphenyl]-2-carbonitrile
Synonyms: 4'-methyl-[1,1'-biphenyl]-2-carbonitrile
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: ZGQVZLSNEBEHFN-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC=CC=C1C#N
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)