missing translation for 'onlineSavingsMsg'
Learn More

2-Cinnamoylthiophene 98.0+%, TCI America™

Catalog No. C07551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07551G 1 g
C07555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07551G Supplier TCI America Supplier No. C07551G
Only null left

Chemical Identifiers

CAS 3988-77-0
Molecular Formula C13H10OS
Molecular Weight (g/mol) 214.282
MDL Number MFCD00059742
InChI Key DDNPADUKGZMCHV-CMDGGOBGSA-N
Synonym 3-Phenyl-1-(2-thienyl)-2-propen-1-one, Styryl 2-Thienyl Ketone
PubChem CID 5702653
IUPAC Name (E)-3-phenyl-1-thiophen-2-ylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C=CC(=O)C2=CC=CS2

Specifications

Melting Point 82°C
Color Yellow
Quantity 1 g
Formula Weight 214.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cinnamoylthiophene