missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorothioanisole 98.0+%, TCI America™
$41.02 - $115.57
Chemical Identifiers
| CAS | 17733-22-1 |
|---|---|
| Molecular Formula | C7H7ClS |
| Molecular Weight (g/mol) | 158.64 |
| MDL Number | MFCD00040124 |
| InChI Key | IHLDFHCSSCVPQW-UHFFFAOYSA-N |
| Synonym | 2-Chlorophenyl Methyl Sulfide, 1-Chloro-2-(methylthio)benzene |
| PubChem CID | 2737363 |
| IUPAC Name | 1-chloro-2-(methylsulfanyl)benzene |
| SMILES | CSC1=CC=CC=C1Cl |
Chemical Identifiers
| 17733-22-1 | |
| 158.64 | |
| IHLDFHCSSCVPQW-UHFFFAOYSA-N | |
| 2737363 | |
| CSC1=CC=CC=C1Cl |
| C7H7ClS | |
| MFCD00040124 | |
| 2-Chlorophenyl Methyl Sulfide, 1-Chloro-2-(methylthio)benzene | |
| 1-chloro-2-(methylsulfanyl)benzene |
Specifications
| 17733-22-1 | |
| C7H7ClS | |
| 5 g | |
| IHLDFHCSSCVPQW-UHFFFAOYSA-N | |
| 1-chloro-2-(methylsulfanyl)benzene | |
| 2737363 | |
| ≥98.0% (GC) | |
| 2-Chlorothioanisole |
| 227°C | |
| MFCD00040124 | |
| 2-Chlorophenyl Methyl Sulfide, 1-Chloro-2-(methylthio)benzene | |
| CSC1=CC=CC=C1Cl | |
| 158.64 | |
| 158.64 | |
| Liquid |
Safety and Handling
TSCA : No