missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorothioanisole 98.0+%, TCI America™
Supplier: TCI America C206925G
Specifications
| 2-Chlorothioanisole | |
| 227°C | |
| MFCD00040124 | |
| 2-Chlorophenyl Methyl Sulfide, 1-Chloro-2-(methylthio)benzene | |
| CSC1=CC=CC=C1Cl | |
| 158.64 | |
| 158.64 | |
| Liquid |
| 17733-22-1 | |
| C7H7ClS | |
| 25 g | |
| IHLDFHCSSCVPQW-UHFFFAOYSA-N | |
| 1-chloro-2-(methylsulfanyl)benzene | |
| 2737363 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 17733-22-1 | |
| 158.64 | |
| IHLDFHCSSCVPQW-UHFFFAOYSA-N | |
| 2737363 | |
| CSC1=CC=CC=C1Cl |
| C7H7ClS | |
| MFCD00040124 | |
| 2-Chlorophenyl Methyl Sulfide, 1-Chloro-2-(methylthio)benzene | |
| 1-chloro-2-(methylsulfanyl)benzene |
Safety and Handling
TSCA : No