missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorostyrene (stabilized with TBC) 96.0+%, TCI America™
Supplier: TCI America C02885G
Specifications
| 2-Chlorostyrene (stabilized with TBC) | |
| -63°C | |
| 187°C | |
| MFCD00000567 | |
| 1993 | |
| ISRGONDNXBCDBM-UHFFFAOYSA-N | |
| 1-chloro-2-ethenylbenzene | |
| 14906 | |
| ≥96.0% (GC) |
| 2039-87-4 | |
| Yellow | |
| C8H7Cl | |
| 5 g | |
| 2-chlorostyrene, 1-chloro-2-vinylbenzene, o-chlorostyrene, benzene, 1-chloro-2-ethenyl, styrene, o-chloro, ortho-chlorostyrene, stryene, o-chloro, styrene, 2-chloro, unii-za43r4q315, ccris 4598 | |
| C=CC1=CC=CC=C1Cl | |
| 138.594 | |
| 138.59 | |
| Liquid |
Chemical Identifiers
| 2039-87-4 | |
| 138.594 | |
| ISRGONDNXBCDBM-UHFFFAOYSA-N | |
| 14906 | |
| C=CC1=CC=CC=C1Cl |
| C8H7Cl | |
| MFCD00000567 | |
| 2-chlorostyrene, 1-chloro-2-vinylbenzene, o-chlorostyrene, benzene, 1-chloro-2-ethenyl, styrene, o-chloro, ortho-chlorostyrene, stryene, o-chloro, styrene, 2-chloro, unii-za43r4q315, ccris 4598 | |
| 1-chloro-2-ethenylbenzene |
Safety and Handling
EINECSNumber : (3)-3441
RTECSNumber : WL4160000
TSCA : No
Recommended Storage : Refrigerator