missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorostyrene (stabilized with TBC) 96.0+%, TCI America™

Catalog No. p-7124338
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C02885G 5 g
C028825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C02885G Supplier TCI America Supplier No. C02885G

Chemical Identifiers

CAS 2039-87-4
Molecular Formula C8H7Cl
Molecular Weight (g/mol) 138.594
MDL Number MFCD00000567
InChI Key ISRGONDNXBCDBM-UHFFFAOYSA-N
Synonym 2-chlorostyrene, 1-chloro-2-vinylbenzene, o-chlorostyrene, benzene, 1-chloro-2-ethenyl, styrene, o-chloro, ortho-chlorostyrene, stryene, o-chloro, styrene, 2-chloro, unii-za43r4q315, ccris 4598
PubChem CID 14906
IUPAC Name 1-chloro-2-ethenylbenzene
SMILES C=CC1=CC=CC=C1Cl

Specifications

Melting Point -63°C
Color Yellow
Boiling Point 187°C
Quantity 5 g
UN Number 1993
Formula Weight 138.59
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorostyrene (stabilized with TBC)
EINECSNumber (3)-3441
RTECSNumber WL4160000
TSCA No
Recommended Storage Refrigerator