missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorophenylhydrazine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America C082625G
Specifications
| 2-Chlorophenylhydrazine Hydrochloride | |
| C6H8Cl2N2 | |
| 25 g | |
| ADODRSVGNHNKAT-UHFFFAOYSA-N | |
| hydrogen (2-chlorophenyl)hydrazine chloride | |
| 443287 | |
| 179.04 | |
| Crystalline Powder |
| 41052-75-9 | |
| MFCD00012928 | |
| 2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride | |
| [H+].[Cl-].NNC1=CC=CC=C1Cl | |
| 179.04 | |
| CHEBI:1046 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 41052-75-9 | |
| 179.04 | |
| ADODRSVGNHNKAT-UHFFFAOYSA-N | |
| 443287 | |
| hydrogen (2-chlorophenyl)hydrazine chloride |
| C6H8Cl2N2 | |
| MFCD00012928 | |
| 2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride | |
| CHEBI:1046 | |
| [H+].[Cl-].NNC1=CC=CC=C1Cl |
Safety and Handling
EINECSNumber : (3)-0471
TSCA : No