missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7124632
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C082625G 25 g
C0826100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C082625G Supplier TCI America Supplier No. C082625G
Only null left

Chemical Identifiers

CAS 41052-75-9
Molecular Formula C6H8Cl2N2
Molecular Weight (g/mol) 179.04
MDL Number MFCD00012928
InChI Key ADODRSVGNHNKAT-UHFFFAOYSA-N
Synonym 2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride
PubChem CID 443287
ChEBI CHEBI:1046
IUPAC Name hydrogen (2-chlorophenyl)hydrazine chloride
SMILES [H+].[Cl-].NNC1=CC=CC=C1Cl

Specifications

Quantity 25 g
Formula Weight 179.04
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorophenylhydrazine Hydrochloride
EINECSNumber (3)-0471
TSCA No