Learn More
2-Chlorophenylacetonitrile, 98+%
CAS: 2856-63-5 | C8H6ClN | 151.593 g/mol
Supplier: Thermo Scientific Chemicals A1317618
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Chlorophenylacetonitrile | |
| 21°C to 24°C | |
| >110°C (230°F) | |
| 1.544 | |
| 50 g | |
| 2043683 | |
| MRDUURPIPLIGQX-UHFFFAOYSA-N | |
| 2-(2-chlorophenyl)acetonitrile | |
| 76112 | |
| ≥98% |
| 2856-63-5 | |
| 240°C to 242°C | |
| C8H6ClN | |
| MFCD00001898 | |
| UN3439 | |
| 2-chlorobenzyl cyanide, 2-chlorophenylacetonitrile, 2-2-chlorophenyl acetonitrile, o-chlorobenzyl cyanide, benzeneacetonitrile, 2-chloro, 2-chlorobenzeneacetonitrile, 2-chlorobenzylcyanide, 2-chlorophenyl acetonitrile, o-chlorophenyl acetonitrile, acetonitrile, o-chlorophenyl | |
| C1=CC=C(C(=C1)CC#N)Cl | |
| 151.593 | |
| 151.6 |
Chemical Identifiers
| 2856-63-5 | |
| 151.593 | |
| MRDUURPIPLIGQX-UHFFFAOYSA-N | |
| 76112 | |
| C1=CC=C(C(=C1)CC#N)Cl |
| C8H6ClN | |
| MFCD00001898 | |
| 2-chlorobenzyl cyanide, 2-chlorophenylacetonitrile, 2-2-chlorophenyl acetonitrile, o-chlorobenzyl cyanide, benzeneacetonitrile, 2-chloro, 2-chlorobenzeneacetonitrile, 2-chlorobenzylcyanide, 2-chlorophenyl acetonitrile, o-chlorophenyl acetonitrile, acetonitrile, o-chlorophenyl | |
| 2-(2-chlorophenyl)acetonitrile |
Safety and Handling
GHS H Statement
H300-H311-H331-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 220-669-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only