missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenylacetone, 96%

Catalog No. AAL0984603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0984603 1 g
AAL0984606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0984603 Supplier Thermo Scientific Chemicals Supplier No. L0984603

CAS: 6305-95-9 | C9H9ClO | 168.62 g/mol

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Difficult to mix in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.

Chemical Identifiers

CAS 6305-95-9
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00045175
InChI Key LWGNDIMNCPMZOF-UHFFFAOYSA-N
Synonym 2-chlorophenylacetone, 1-2-chlorophenyl propan-2-one, 1-2-chlorophenyl acetone, 2-propanone, 1-2-chlorophenyl, acmc-20apcv, 2-chlorphenyl acetone, 1-2-chlorophenyl acetone #, 1-2-chlorophenyl-2-propanone, 1-2-chloro-phenyl-propan-2-one
PubChem CID 229355
IUPAC Name 1-(2-chlorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC=C1Cl

Specifications

Boiling Point 102°C to 103°C (3 mmHg)
Refractive Index 1.536
Quantity 1 g
Beilstein 2043542
Solubility Information Difficult to mix in water.
Formula Weight 168.62
Percent Purity 96%
Chemical Name or Material 2-Chlorophenylacetone
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only