missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl phosphorodichloridate, 98+%

Catalog No. p-7081502
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1150506 5 g
AAA1150514 25 g
AAA1150522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1150506 Supplier Thermo Scientific Chemicals Supplier No. A1150506

CAS: 15074-54-1 | C6H4Cl3O2P | 245.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15074-54-1
Molecular Formula C6H4Cl3O2P
Molecular Weight (g/mol) 245.42
MDL Number MFCD00009704
InChI Key VLDPXPPHXDGHEW-UHFFFAOYSA-N
Synonym 2-chlorophenyl phosphorodichloridate, 2-chlorophenyl dichlorophosphate, o-chlorophenyl dichlorophosphate, phosphorodichloridic acid, 2-chlorophenyl ester, o-chlorophenyl phosphorodichloridate, 2-chlorophenyl phosphoric acid dichloride, 2-chlorophenyl chlorophosphonochloridate, acmc-20anxn, o-chlorophenylphosphonyl dichloride, 2-chlorophenyl dichloridophosphate #
PubChem CID 84775
IUPAC Name 1-chloro-2-dichlorophosphoryloxybenzene
SMILES C1=CC=C(C(=C1)OP(=O)(Cl)Cl)Cl

Specifications

Density 1.491
Boiling Point 82°C to 84°C (0.4 mmHg)
Flash Point 105°C (221°F)
Refractive Index 1.538
Quantity 5 g
UN Number UN3265
Beilstein 911009
Sensitivity Moisture sensitive
Formula Weight 245.43
Percent Purity ≥98%
Chemical Name or Material 2-Chlorophenyl phosphorodichloridate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 239-129-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only