accessibility menu, dialog, popup

UserName

CAS RN 15074-54-1

IUPAC Name: 2-chlorophenyl chlorophosphonochloridate
Synonyms: 2-chlorophenyl chlorophosphonochloridate
Molecular Weight (g/mol): 245.42
Molecular Formula: C6H4Cl3O2P
InChi Key: VLDPXPPHXDGHEW-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1OP(Cl)(Cl)=O
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)