missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenol 99.0+%, TCI America™

Catalog No. p-7124305
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C024125G 25 g
C0241500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C024125G Supplier TCI America Supplier No. C024125G
Only null left

Chemical Identifiers

CAS 95-57-8
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.555
MDL Number MFCD00002159
InChI Key ISPYQTSUDJAMAB-UHFFFAOYSA-N
Synonym o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol
PubChem CID 7245
ChEBI CHEBI:47083
IUPAC Name 2-chlorophenol
SMILES C1=CC=C(C(=C1)O)Cl

Specifications

Color Colorless
Boiling Point 175°C
Quantity 25 g
UN Number 2021
Formula Weight 128.56
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chlorophenol
EINECSNumber (3)-0895
RTECSNumber SK2625000
TSCA Yes