Learn More
2-Chlorophenol, 99%
CAS: 95-57-8 | C6H5ClO | 128.555 g/mol
Supplier: Thermo Scientific Chemicals A103110I
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Chlorophenol | |
| 7°C to 8°C | |
| 175°C to 176°C | |
| Phenol-like | |
| 1.559 | |
| 5000 g | |
| 1905114 | |
| o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol | |
| C1=CC=C(C(=C1)O)Cl | |
| 128.555 | |
| CHEBI:47083 | |
| 99% |
| 95-57-8 | |
| 1.26 | |
| 63°C (145°F) | |
| C6H5ClO | |
| MFCD00002159 | |
| UN2021 | |
| 14,2154 | |
| ISPYQTSUDJAMAB-UHFFFAOYSA-N | |
| 2-chlorophenol | |
| 7245 | |
| 128.56 |
Chemical Identifiers
| 95-57-8 | |
| 128.555 | |
| ISPYQTSUDJAMAB-UHFFFAOYSA-N | |
| 7245 | |
| 2-chlorophenol |
| C6H5ClO | |
| MFCD00002159 | |
| o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol | |
| CHEBI:47083 | |
| C1=CC=C(C(=C1)O)Cl |
Safety and Handling
GHS H Statement
H302-H312-H332-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
P210-P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
H227-H300-H312+H332-H315-H317-H318
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, LIQUID
EINECSNumber : 202-433-2
RTECSNumber : SK2625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only