missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenol, 98+%

Catalog No. p-90437
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC180990050 5 g Glass bottle
AC180991000 100 g Glass bottle
AC180995000 500 g Glass bottle
AC180990025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC180990050 Supplier Thermo Scientific Chemicals Supplier No. 180990050

CAS: 95-57-8 | C6H5ClO | 128.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-57-8
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.56
MDL Number MFCD00002159
InChI Key ISPYQTSUDJAMAB-UHFFFAOYSA-N
Synonym o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol
PubChem CID 7245
ChEBI CHEBI:47083
IUPAC Name 2-chlorophenol
SMILES C1=CC=C(C(=C1)O)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 8.0°C
Density 1.2620g/mL
Boiling Point 173.0°C to 175.0°C
Flash Point 63°C
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5580 to 1.5600
Linear Formula ClC6H4OH
Quantity 5 g
Beilstein 06,183
Merck Index 15,2157
Specific Gravity 1.262
Solubility Information Solubility in water: 28.5g/l (20°C). Other solubilities: freely soluble in solutions of sodium hydroxide,, alcohol and ether, soluble in aromatic solvents
Viscosity 4.11 mPa.s (25°C)
Formula Weight 128.56
Percent Purity 98+%
Chemical Name or Material 2-Chlorophenol, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa
EINECSNumber 202-433-2
RTECSNumber SK2625000
TSCA TSCA

RUO – Research Use Only