missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 33575-83-6 |
|---|---|
| Molecular Formula | C9H7ClN2O |
| Molecular Weight (g/mol) | 194.62 |
| MDL Number | MFCD00466332 |
| InChI Key | AGLNTFQAHIRTFA-UHFFFAOYSA-N |
| Synonym | 2-chloromethyl-5-phenyl-1,3,4-oxadiazole, 2-chloromethyl-5-phenyl-1,3,4 oxadiazole, 5-chloromethyl-2-phenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-chloromethyl-5-phenyl, enamine_005035, chloromethylphenyloxadiazole, 1,3,4-oxadiazole,2-chloromethyl-5-phenyl, 5-chloromethyl-1,3,4-oxadiazol-2-yl benzene |
| PubChem CID | 314941 |
| IUPAC Name | 2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole |
| SMILES | ClCC1=NN=C(O1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC438280010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 33575-83-6 | |
| 194.62 | |
| AGLNTFQAHIRTFA-UHFFFAOYSA-N | |
| 314941 | |
| ClCC1=NN=C(O1)C1=CC=CC=C1 |
| C9H7ClN2O | |
| MFCD00466332 | |
| 2-chloromethyl-5-phenyl-1,3,4-oxadiazole, 2-chloromethyl-5-phenyl-1,3,4 oxadiazole, 5-chloromethyl-2-phenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-chloromethyl-5-phenyl, enamine_005035, chloromethylphenyloxadiazole, 1,3,4-oxadiazole,2-chloromethyl-5-phenyl, 5-chloromethyl-1,3,4-oxadiazol-2-yl benzene | |
| 2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole |
Specifications
| 33575-83-6 | |
| 100.0 | |
| 96% min. (GC) | |
| C9H7ClN2O | |
| 2-chloromethyl-5-phenyl-1,3,4-oxadiazole, 2-chloromethyl-5-phenyl-1,3,4 oxadiazole, 5-chloromethyl-2-phenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-chloromethyl-5-phenyl, enamine_005035, chloromethylphenyloxadiazole, 1,3,4-oxadiazole,2-chloromethyl-5-phenyl, 5-chloromethyl-1,3,4-oxadiazol-2-yl benzene | |
| ClCC1=NN=C(O1)C1=CC=CC=C1 | |
| 194.62 | |
| 194.62 | |
| 2-(chloromethyl)-5-phenyl-1, 3, 4-oxadiazole |
| 96.0 | |
| Authentic | |
| Glass Bottle | |
| MFCD00466332 | |
| AGLNTFQAHIRTFA-UHFFFAOYSA-N | |
| 2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole | |
| 314941 | |
| 97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
RUO – Research Use Only