CAS RN 33575-83-6
IUPAC Name:
2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole
Synonyms:
2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole
Molecular Weight (g/mol):
194.62
Molecular Formula:
C9H7ClN2O
InChi Key:
AGLNTFQAHIRTFA-UHFFFAOYSA-N
SMILES:
ClCC1=NN=C(O1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole, 97%, Thermo Scientific™
2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole, 97%, Thermo Scientific™
CAS: 33575-83-6 Molecular Formula: C9H7ClN2O Molecular Weight (g/mol): 194.62 MDL Number: MFCD00466332 InChI Key: AGLNTFQAHIRTFA-UHFFFAOYSA-N Synonym: 2-chloromethyl-5-phenyl-1,3,4-oxadiazole,2-chloromethyl-5-phenyl-1,3,4 oxadiazole,5-chloromethyl-2-phenyl-1,3,4-oxadiazole,1,3,4-oxadiazole, 2-chloromethyl-5-phenyl,enamine_005035,chloromethylphenyloxadiazole,1,3,4-oxadiazole,2-chloromethyl-5-phenyl,5-chloromethyl-1,3,4-oxadiazol-2-yl benzene PubChem CID: 314941 IUPAC Name: 2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole SMILES: ClCC1=NN=C(O1)C1=CC=CC=C1