missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 438280010

Quantity 1g

 View more versions of this product

Catalog No. AC438280010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

33575-83-6
194.62
AGLNTFQAHIRTFA-UHFFFAOYSA-N
314941
ClCC1=NN=C(O1)C1=CC=CC=C1
C9H7ClN2O
MFCD00466332
2-chloromethyl-5-phenyl-1,3,4-oxadiazole, 2-chloromethyl-5-phenyl-1,3,4 oxadiazole, 5-chloromethyl-2-phenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-chloromethyl-5-phenyl, enamine_005035, chloromethylphenyloxadiazole, 1,3,4-oxadiazole,2-chloromethyl-5-phenyl, 5-chloromethyl-1,3,4-oxadiazol-2-yl benzene
2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole

Specifications

2-(chloromethyl)-5-phenyl-1, 3, 4-oxadiazole
96.0
Authentic
Glass Bottle
MFCD00466332
2-chloromethyl-5-phenyl-1,3,4-oxadiazole, 2-chloromethyl-5-phenyl-1,3,4 oxadiazole, 5-chloromethyl-2-phenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-chloromethyl-5-phenyl, enamine_005035, chloromethylphenyloxadiazole, 1,3,4-oxadiazole,2-chloromethyl-5-phenyl, 5-chloromethyl-1,3,4-oxadiazol-2-yl benzene
ClCC1=NN=C(O1)C1=CC=CC=C1
194.62
194.62
33575-83-6
100.0
96% min. (GC)
C9H7ClN2O
1g
AGLNTFQAHIRTFA-UHFFFAOYSA-N
2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole
314941
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...