missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Chloromethyl)-4-methylquinazoline 98.0+%, TCI America™
Supplier: TCI America C29281G
Specifications
| 2-(Chloromethyl)-4-methylquinazoline | |
| 63°C | |
| MFCD09807547 | |
| 3261 | |
| CC1=C2C=CC=CC2=NC(CCl)=N1 | |
| 192.65 | |
| 192.65 | |
| Crystalline Powder |
| 109113-72-6 | |
| C10H9ClN2 | |
| 1 g | |
| UHCUBOJGMLASBY-UHFFFAOYSA-N | |
| 2-(chloromethyl)-4-methylquinazoline | |
| 241518 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 109113-72-6 | |
| 192.65 | |
| UHCUBOJGMLASBY-UHFFFAOYSA-N | |
| 2-(chloromethyl)-4-methylquinazoline |
| C10H9ClN2 | |
| MFCD09807547 | |
| 241518 | |
| CC1=C2C=CC=CC2=NC(CCl)=N1 |
Safety and Handling
TSCA : No