missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-4-methylquinazoline 98.0+%, TCI America™

Catalog No. p-7125727
Change view
Click to view available options
Quantity:
200 mg
1 g
Catalog No. Quantity
C2928200MG 200 mg
C29281G 1 g
2 options

Catalog No. C2928200MG

Supplier: TCI America C2928200MG

Only null left

Chemical Identifiers

CAS 109113-72-6
Molecular Formula C10H9ClN2
Molecular Weight (g/mol) 192.65
MDL Number MFCD09807547
InChI Key UHCUBOJGMLASBY-UHFFFAOYSA-N
PubChem CID 241518
IUPAC Name 2-(chloromethyl)-4-methylquinazoline
SMILES CC1=C2C=CC=CC2=NC(CCl)=N1

Specifications

Melting Point 63°C
Quantity 200 mg
UN Number 3261
Formula Weight 192.65
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Chloromethyl)-4-methylquinazoline