accessibility menu, dialog, popup

UserName

CH3NNCl

CAS RN 109113-72-6

IUPAC Name: 2-(chloromethyl)-4-methylquinazoline
Synonyms: 2-(chloromethyl)-4-methylquinazoline
Molecular Weight (g/mol): 192.65
Molecular Formula: C10H9ClN2
InChi Key: UHCUBOJGMLASBY-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=NC(CCl)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)