missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl phenyl sulfone, 98%

Catalog No. p-7025946
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2169014 25 g
AAB2169022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2169014 Supplier Thermo Scientific Chemicals Supplier No. B2169014

CAS: 938-09-0 | C8H9ClO2S | 204.668 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 938-09-0
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD00025047
InChI Key NUJGORANFDSMOL-UHFFFAOYSA-N
Synonym 2-chloroethyl phenyl sulfone, 2-chloroethanesulfonyl benzene, 2-chloroethyl sulfonyl benzene, 2-chloroethylphenylsulfone, 2-chloroethylsulfonyl benzene, sulfone, 2-chloroethyl phenyl, 2-chloroethyl sulphonyl benzene, benzene, 2-chloroethyl sulfonyl, 2-chloroethylphenyl sulfone
PubChem CID 13646
IUPAC Name 2-chloroethylsulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)CCCl

Specifications

Melting Point 52°C to 55°C
Boiling Point 168°C to 174°C (7 mmHg)
Quantity 25 g
UN Number UN1759
Beilstein 1104341
Formula Weight 204.68
Percent Purity 98%
Chemical Name or Material 2-Chloroethyl phenyl sulfone
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 213-337-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only