Learn More
2-Chlorobenzylamine, 96%
CAS: 89-97-4 | C7H8ClN | 141.598 g/mol
Supplier: Thermo Scientific Chemicals A1559422
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Chlorobenzylamine | |
| 1.17 | |
| 88°C (190°F) | |
| C7H8ClN | |
| MFCD00008108 | |
| UN2735 | |
| Air Sensitive | |
| KDDNKZCVYQDGKE-UHFFFAOYSA-N | |
| (2-chlorophenyl)methanamine | |
| 66648 | |
| 96% |
| 89-97-4 | |
| 227°C | |
| Amine-like | |
| 1.562 | |
| 100 g | |
| 907186 | |
| 2-chlorobenzylamine, 2-chlorophenyl methanamine, o-chlorobenzylamine, benzenemethanamine, 2-chloro, o-chlorobenzyl amine, 2-chlorobenzenemethanamine, 2-chloro-benzylamine, benzylamine, o-chloro, 2-chlorophenyl methylamine, unii-l790j4y7a3 | |
| C1=CC=C(C(=C1)CN)Cl | |
| 141.598 | |
| 141.6 |
Chemical Identifiers
| 89-97-4 | |
| 141.598 | |
| KDDNKZCVYQDGKE-UHFFFAOYSA-N | |
| 66648 | |
| C1=CC=C(C(=C1)CN)Cl |
| C7H8ClN | |
| MFCD00008108 | |
| 2-chlorobenzylamine, 2-chlorophenyl methanamine, o-chlorobenzylamine, benzenemethanamine, 2-chloro, o-chlorobenzyl amine, 2-chlorobenzenemethanamine, 2-chloro-benzylamine, benzylamine, o-chloro, 2-chlorophenyl methylamine, unii-l790j4y7a3 | |
| (2-chlorophenyl)methanamine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 201-955-8
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only