accessibility menu, dialog, popup

UserName

H2NCl

CAS RN 89-97-4

IUPAC Name: 1-(2-chlorophenyl)methanamine
Synonyms: 1-(2-chlorophenyl)methanamine
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: KDDNKZCVYQDGKE-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1Cl
Boiling Point 
  • (2)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)