missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzylamine, 96%

Catalog No. AAA1559414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1559414 25 g
AAA1559422 100 g
AAA1559436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1559414 Supplier Thermo Scientific Chemicals Supplier No. A1559414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 89-97-4 | C7H8ClN | 141.598 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-97-4
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00008108
InChI Key KDDNKZCVYQDGKE-UHFFFAOYSA-N
Synonym 2-chlorobenzylamine, 2-chlorophenyl methanamine, o-chlorobenzylamine, benzenemethanamine, 2-chloro, o-chlorobenzyl amine, 2-chlorobenzenemethanamine, 2-chloro-benzylamine, benzylamine, o-chloro, 2-chlorophenyl methylamine, unii-l790j4y7a3
PubChem CID 66648
IUPAC Name (2-chlorophenyl)methanamine
SMILES C1=CC=C(C(=C1)CN)Cl

Specifications

Density 1.17
Boiling Point 227°C
Flash Point 88°C (190°F)
Odor Amine-like
Refractive Index 1.562
Quantity 25 g
UN Number UN2735
Beilstein 907186
Sensitivity Air Sensitive
Formula Weight 141.6
Percent Purity 96%
Chemical Name or Material 2-Chlorobenzylamine
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 201-955-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only