missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl Alcohol 98.0+%, TCI America™

Catalog No. C097325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C097325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C097325G Supplier TCI America Supplier No. C097325G

Chemical Identifiers

CAS 17849-38-6
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00004604
InChI Key MBYQPPXEXWRMQC-UHFFFAOYSA-N
Synonym 2-chlorobenzyl alcohol, 2-chlorophenyl methanol, benzenemethanol, 2-chloro, 2-chlorobenzenemethanol, o-chlorobenzyl alcohol, benzyl alcohol, o-chloro, 2-chloro-phenyl-methanol, 2-chlorophenyl methan-1-ol, benzenemethanol, ar-chloro, 2-chlorobenzylalcohol
PubChem CID 28810
IUPAC Name (2-chlorophenyl)methanol
SMILES C1=CC=C(C(=C1)CO)Cl

Specifications

Melting Point 71°C
Color White-Yellow
Boiling Point 135°C
Quantity 25 g
Formula Weight 142.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorobenzyl Alcohol
EINECSNumber (3)-1050
TSCA No