accessibility menu, dialog, popup

UserName

HOCl

CAS RN 17849-38-6

IUPAC Name: (2-chlorophenyl)methanol
Synonyms: benzenemethanol, 2-chloro-
Molecular Weight (g/mol): 142.58
Molecular Formula: C7H7ClO
InChi Key: MBYQPPXEXWRMQC-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1Cl
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)