missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl alcohol, 99%

Catalog No. AC163420100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC163420100 10 g
AC163420500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163420100 Supplier Thermo Scientific Chemicals Supplier No. 163420100

CAS: 17849-38-6 | C7H7ClO | 142.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17849-38-6
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00004604
InChI Key MBYQPPXEXWRMQC-UHFFFAOYSA-N
Synonym 2-chlorobenzyl alcohol, 2-chlorophenyl methanol, benzenemethanol, 2-chloro, 2-chlorobenzenemethanol, o-chlorobenzyl alcohol, benzyl alcohol, o-chloro, 2-chloro-phenyl-methanol, 2-chlorophenyl methan-1-ol, benzenemethanol, ar-chloro, 2-chlorobenzylalcohol
PubChem CID 28810
IUPAC Name (2-chlorophenyl)methanol
SMILES C1=CC=C(C(=C1)CO)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 69°C to 72°C
Color White
Boiling Point 227°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CH2OH
Quantity 10 g
Beilstein 06, 444
Solubility Information Solubility in water: limited soluble in water. Other solubilities: 1g/10mL in methanol
Formula Weight 142.58
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Chlorobenzyl alcohol, 99%
Show More Show Less
EINECSNumber 241-801-7
TSCA TSCA

RUO – Research Use Only