missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7124241
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
C014025ML 25 mL
C0140100ML 100 mL
C0140500ML 500 mL
3 options

Catalog No. C014025ML

Supplier: TCI America C014025ML

Only null left

Chemical Identifiers

CAS 609-65-4
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00000660
InChI Key ONIKNECPXCLUHT-UHFFFAOYSA-N
Synonym benzoyl chloride, 2-chloro, o-chlorobenzoyl chloride, chlorobenzoyl chloride, benzoyl chloride, o-chloro, ococ, 2-chloro-benzoyl chloride, unii-jr29a4n74x, o-chlorobenzoylchloride, 2-chlorobenzoylchloride, 2-chlorobenzoic acid chloride
PubChem CID 69110
ChEBI CHEBI:60719
IUPAC Name 2-chlorobenzoyl chloride
SMILES ClC(=O)C1=CC=CC=C1Cl

Specifications

Color Yellow
Boiling Point 230°C
Quantity 25 mL
UN Number 3265
Formula Weight 175.01
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzoyl Chloride
EINECSNumber (3)-0062&(3)-1409
TSCA Yes