missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. C0140500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0140500ML 500 mL
C014025ML 25 mL
C0140100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0140500ML Supplier TCI America Supplier No. C0140500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 609-65-4
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00000660
InChI Key ONIKNECPXCLUHT-UHFFFAOYSA-N
Synonym benzoyl chloride, 2-chloro, o-chlorobenzoyl chloride, chlorobenzoyl chloride, benzoyl chloride, o-chloro, ococ, 2-chloro-benzoyl chloride, unii-jr29a4n74x, o-chlorobenzoylchloride, 2-chlorobenzoylchloride, 2-chlorobenzoic acid chloride
PubChem CID 69110
ChEBI CHEBI:60719
IUPAC Name 2-chlorobenzoyl chloride
SMILES ClC(=O)C1=CC=CC=C1Cl

Specifications

Color Yellow
Boiling Point 230°C
Quantity 500 mL
UN Number 3265
Formula Weight 175.01
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzoyl Chloride
EINECSNumber (3)-0062&(3)-1409
TSCA Yes