missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoyl chloride, 97%

Catalog No. p-7022153
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1478522 100 g
AAA1478530 250 g
AAA147850B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1478522 Supplier Thermo Scientific Chemicals Supplier No. A1478522
Only null left

CAS: 609-65-4 | C7H4Cl2O | 175.01 g/mol

2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.

Solubility
Soluble in acetone, ether, and alcohol. Solubility in water it decomposes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong bases, alcohols, oxidizing agents.

Chemical Identifiers

CAS 609-65-4
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00000660
InChI Key ONIKNECPXCLUHT-UHFFFAOYSA-N
Synonym benzoyl chloride, 2-chloro, o-chlorobenzoyl chloride, chlorobenzoyl chloride, benzoyl chloride, o-chloro, ococ, 2-chloro-benzoyl chloride, unii-jr29a4n74x, o-chlorobenzoylchloride, 2-chlorobenzoylchloride, 2-chlorobenzoic acid chloride
PubChem CID 69110
ChEBI CHEBI:60719
IUPAC Name 2-chlorobenzoyl chloride
SMILES ClC(=O)C1=CC=CC=C1Cl

Specifications

Melting Point -4°C
Density 1.379
Boiling Point 237°C to 239°C
Flash Point 125°C (257°F)
Odor Acrid
Refractive Index 1.5718
Quantity 100 g
UN Number UN3265
Beilstein 386435
Sensitivity Moisture sensitive
Solubility Information Soluble in acetone,ether,and alcohol. Solubility in water it decomposes.
Formula Weight 175.01
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzoyl chloride
Show More Show Less
Hazard Category H290-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 210-194-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only