missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzoxazole 98.0+%, TCI America™
Supplier: TCI America C13875G
Specifications
| 2-Chlorobenzoxazole | |
| 7°C | |
| 202°C | |
| MFCD00005766 | |
| 2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole | |
| ClC1=NC2=CC=CC=C2O1 | |
| 153.57 | |
| 153.57 | |
| Liquid |
| 615-18-9 | |
| Yellow | |
| C7H4ClNO | |
| 5 g | |
| BBVQDWDBTWSGHQ-UHFFFAOYSA-N | |
| 2-chloro-1,3-benzoxazole | |
| 11986 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 615-18-9 | |
| 153.57 | |
| BBVQDWDBTWSGHQ-UHFFFAOYSA-N | |
| 11986 | |
| ClC1=NC2=CC=CC=C2O1 |
| C7H4ClNO | |
| MFCD00005766 | |
| 2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole | |
| 2-chloro-1,3-benzoxazole |
Safety and Handling
RTECSNumber : DM4680000
TSCA : Yes