missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoxazole 98.0+%, TCI America™

Catalog No. p-7124949
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13875G 5 g
C138725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13875G Supplier TCI America Supplier No. C13875G
Only null left

Chemical Identifiers

CAS 615-18-9
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.57
MDL Number MFCD00005766
InChI Key BBVQDWDBTWSGHQ-UHFFFAOYSA-N
Synonym 2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole
PubChem CID 11986
IUPAC Name 2-chloro-1,3-benzoxazole
SMILES ClC1=NC2=CC=CC=C2O1

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 202°C
Quantity 5 g
Formula Weight 153.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzoxazole
RTECSNumber DM4680000
TSCA Yes