accessibility menu, dialog, popup

UserName

CAS RN 615-18-9

IUPAC Name: 2-chloro-1,3-benzoxazole
Synonyms: benzoxazole, 2-chloro-
Molecular Weight (g/mol): 153.57
Molecular Formula: C7H4ClNO
InChi Key: BBVQDWDBTWSGHQ-UHFFFAOYSA-N
SMILES: ClC1=NC2=CC=CC=C2O1
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)