Learn More
2-Chlorobenzoxazole, 98%
CAS: 615-18-9 | C7H4ClNO | 153.57 g/mol
Supplier: Thermo Scientific Chemicals L0798114
Description
2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Not miscible in water.
Notes
Store under inert gas. Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Keep away from heat.
Specifications
| 2-Chlorobenzoxazole | |
| 1.39 | |
| 85°C (185°F) | |
| 1.567 | |
| 25 g | |
| 2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole | |
| BBVQDWDBTWSGHQ-UHFFFAOYSA-N | |
| 2-chloro-1,3-benzoxazole | |
| 11986 | |
| 99% |
| 615-18-9 | |
| 201°C to 202°C | |
| C7H4ClNO | |
| MFCD00005766 | |
| 115982 | |
| Not miscible in water. | |
| ClC1=NC2=CC=CC=C2O1 | |
| 153.57 | |
| 153.57 |
Chemical Identifiers
| 615-18-9 | |
| 153.57 | |
| BBVQDWDBTWSGHQ-UHFFFAOYSA-N | |
| 11986 | |
| ClC1=NC2=CC=CC=C2O1 |
| C7H4ClNO | |
| MFCD00005766 | |
| 2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole | |
| 2-chloro-1,3-benzoxazole |
Safety and Handling
GHS H Statement
H302-H315-H319-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c
H227-H302-H315-H319-H335
EINECSNumber : 210-414-5
RTECSNumber : DM4680000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only