missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoxazole, 98%

Catalog No. p-7044729
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0798106 5 g
AAL0798114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0798106 Supplier Thermo Scientific Chemicals Supplier No. L0798106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 615-18-9 | C7H4ClNO | 153.57 g/mol

2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible in water.

Notes
Store under inert gas. Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Keep away from heat.

Chemical Identifiers

CAS 615-18-9
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.57
MDL Number MFCD00005766
InChI Key BBVQDWDBTWSGHQ-UHFFFAOYSA-N
Synonym 2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole
PubChem CID 11986
IUPAC Name 2-chloro-1,3-benzoxazole
SMILES ClC1=NC2=CC=CC=C2O1

Specifications

Density 1.39
Boiling Point 201°C to 202°C
Flash Point 85°C (185°F)
Refractive Index 1.567
Quantity 5 g
Beilstein 115982
Solubility Information Not miscible in water.
Formula Weight 153.57
Percent Purity 99%
Chemical Name or Material 2-Chlorobenzoxazole
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c
EINECSNumber 210-414-5
RTECSNumber DM4680000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only