Learn More
2-Chlorobenzothiazole, 98%
CAS: 615-20-3 | C7H4ClNS | 169.626 g/mol
$65.32 - $584.32
Chemical Identifiers
| CAS | 615-20-3 |
|---|---|
| Molecular Formula | C7H4ClNS |
| Molecular Weight (g/mol) | 169.626 |
| MDL Number | MFCD00005776 |
| InChI Key | BSQLQMLFTHJVKS-UHFFFAOYSA-N |
| Synonym | 2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole |
| PubChem CID | 11987 |
| IUPAC Name | 2-chloro-1,3-benzothiazole |
| SMILES | C1=CC=C2C(=C1)N=C(S2)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1776206
|
Thermo Scientific Chemicals
A1776206 |
5 g |
Each for $65.32
|
|
|||||
|
AAA1776214
|
Thermo Scientific Chemicals
A1776214 |
25 g |
Each for $206.53
|
|
|||||
|
AAA1776222
|
Thermo Scientific Chemicals
A1776222 |
100 g |
Each for $584.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 615-20-3 | |
| 169.626 | |
| BSQLQMLFTHJVKS-UHFFFAOYSA-N | |
| 11987 | |
| C1=CC=C2C(=C1)N=C(S2)Cl |
| C7H4ClNS | |
| MFCD00005776 | |
| 2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole | |
| 2-chloro-1,3-benzothiazole |
Specifications
| 615-20-3 | |
| 1.37 | |
| >110°C (230°F) | |
| 1.637 | |
| 5 g | |
| 2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole | |
| C1=CC=C2C(=C1)N=C(S2)Cl | |
| 169.626 | |
| 169.63 | |
| 2-Chlorobenzothiazole |
| 30°C to 33°C | |
| 141°C (30 mmHg) | |
| C7H4ClNS | |
| MFCD00005776 | |
| 116316 | |
| BSQLQMLFTHJVKS-UHFFFAOYSA-N | |
| 2-chloro-1,3-benzothiazole | |
| 11987 | |
| ≥98% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 210-415-0
RTECSNumber : DL3150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only