missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzothiazole, 98%

CAS: 615-20-3 | C7H4ClNS | 169.626 g/mol

Supplier:  Thermo Scientific Chemicals A1776214

 View more versions of this product

Catalog No. AAA1776214

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chlorobenzothiazole
30°C to 33°C
141°C (30 mmHg)
C7H4ClNS
MFCD00005776
116316
BSQLQMLFTHJVKS-UHFFFAOYSA-N
2-chloro-1,3-benzothiazole
11987
≥98%
615-20-3
1.37
>110°C (230°F)
1.637
25 g
2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole
C1=CC=C2C(=C1)N=C(S2)Cl
169.626
169.63

Chemical Identifiers

615-20-3
169.626
BSQLQMLFTHJVKS-UHFFFAOYSA-N
11987
C1=CC=C2C(=C1)N=C(S2)Cl
C7H4ClNS
MFCD00005776
2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole
2-chloro-1,3-benzothiazole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 210-415-0

RTECSNumber : DL3150000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only