accessibility menu, dialog, popup

UserName

ClSN

CAS RN 615-20-3

IUPAC Name: 2-chloro-1,3-benzothiazole
Synonyms: benzothiazole, 2-chloro-
Molecular Weight (g/mol): 169.63
Molecular Formula: C7H4ClNS
InChi Key: BSQLQMLFTHJVKS-UHFFFAOYSA-N
SMILES: ClC1=NC2=CC=CC=C2S1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)