missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7124235
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C013325G 25 g
C0133500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C013325G Supplier TCI America Supplier No. C013325G
Only null left

Chemical Identifiers

CAS 118-91-2
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD00002412
InChI Key IKCLCGXPQILATA-UHFFFAOYSA-N
Synonym o-chlorobenzoic acid, benzoic acid, o-chloro, benzoic acid, 2-chloro, 2-cba, kyselina o-chlorbenzoova, chlorobenzoic acid, ocba, benzoic acid, chloro, 2-chloro-benzoic acid, chlorobenzoate
PubChem CID 8374
ChEBI CHEBI:30793
IUPAC Name 2-chlorobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)Cl

Specifications

Melting Point 140°C
Color White
Boiling Point 285°C
Quantity 25 g
Formula Weight 156.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorobenzoic Acid
EINECSNumber (3)-0062&(3)-1424
RTECSNumber DG4976000
TSCA Yes