accessibility menu, dialog, popup

UserName

HOOCl

CAS RN 118-91-2

IUPAC Name: 2-chlorobenzoic acid
Synonyms: chlorobenzoate 2-chorobenzoate kyselina o-chlorbenzoova OCBA 2-cba
Molecular Weight (g/mol): 156.57
Molecular Formula: C7H5ClO2
InChi Key: IKCLCGXPQILATA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1Cl
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)