missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoic acid, 98+%

Catalog No. p-7081034
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1083230 250 g
AAA108320B 1000 g
AAA108320I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1083230 Supplier Thermo Scientific Chemicals Supplier No. A1083230
Only null left

CAS: 118-91-2 | C7H5ClO2 | 156.565 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-91-2
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD00002412
InChI Key IKCLCGXPQILATA-UHFFFAOYSA-N
Synonym o-chlorobenzoic acid, benzoic acid, o-chloro, benzoic acid, 2-chloro, 2-cba, kyselina o-chlorbenzoova, chlorobenzoic acid, ocba, benzoic acid, chloro, 2-chloro-benzoic acid, chlorobenzoate
PubChem CID 8374
ChEBI CHEBI:30793
IUPAC Name 2-chlorobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)Cl

Specifications

Melting Point 138°C to 142°C
Density 1.544
Boiling Point 284°C to 286°C
Flash Point 173°C (343°F)
Quantity 250 g
Beilstein 907340
Merck Index 14,2124
Formula Weight 156.57
Percent Purity ≥98%
Chemical Name or Material 2-Chlorobenzoic acid
Hazard Category H315-H319-H500
Hazard Statement GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 204-285-4
RTECSNumber DG4976000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only