missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzohydrazide 98.0+%, TCI America™

Catalog No. C21165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21165G 5 g
C211625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21165G Supplier TCI America Supplier No. C21165G
Only null left

Chemical Identifiers

CAS 5814-05-1
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.60
MDL Number MFCD00007597
InChI Key KPPNLSKVTKSSTG-UHFFFAOYSA-N
Synonym 2-chlorobenzhydrazide, 2-chlorobenzoic hydrazide, 2-chlorobenzoic acid hydrazide, 2-chlorobenzene-1-carbohydrazide, benzoylhydrazine, o-chloro, benzoic acid, 2-chloro-, hydrazide, 2-chlorobenzenecarbohydrazide, chlorobenzohydrazide, o-chlorobenzhydrazide, 2clphcon2
PubChem CID 79883
IUPAC Name 2-chlorobenzohydrazide
SMILES C1=CC=C(C(=C1)C(=O)NN)Cl

Specifications

Melting Point 118°C
Quantity 5 g
Formula Weight 170.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorobenzohydrazide