accessibility menu, dialog, popup

UserName

NH2HNOCl

CAS RN 5814-05-1

IUPAC Name: 2-chlorobenzohydrazide
Synonyms: benzoylhydrazine, O-chloro
Molecular Weight (g/mol): 170.6
Molecular Formula: C7H7ClN2O
InChi Key: KPPNLSKVTKSSTG-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results