missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzenesulfonamide 98.0+%, TCI America™
$123.03 - $1040.14
Chemical Identifiers
| CAS | 6961-82-6 |
|---|---|
| Molecular Formula | C6H6ClNO2S |
| Molecular Weight (g/mol) | 191.629 |
| InChI Key | JCCBZCMSYUSCFM-UHFFFAOYSA-N |
| Synonym | o-chlorobenzenesulfonamide, 2-chlorobenzene-1-sulfonamide, benzenesulfonamide, 2-chloro, unii-4bt2i28a0c, benzenesulfonamide, o-chloro, 2-chlorobenzenesulphonamide, 2-chloro-benzenesulfonamide, chlorobenzenesulfonamide, o-chlorobenzenesulphonamide, benzenesulfonamide, chloro |
| PubChem CID | 81410 |
| IUPAC Name | 2-chlorobenzenesulfonamide |
| SMILES | C1=CC=C(C(=C1)S(=O)(=O)N)Cl |
Chemical Identifiers
| 6961-82-6 | |
| 191.629 | |
| o-chlorobenzenesulfonamide, 2-chlorobenzene-1-sulfonamide, benzenesulfonamide, 2-chloro, unii-4bt2i28a0c, benzenesulfonamide, o-chloro, 2-chlorobenzenesulphonamide, 2-chloro-benzenesulfonamide, chlorobenzenesulfonamide, o-chlorobenzenesulphonamide, benzenesulfonamide, chloro | |
| 2-chlorobenzenesulfonamide |
| C6H6ClNO2S | |
| JCCBZCMSYUSCFM-UHFFFAOYSA-N | |
| 81410 | |
| C1=CC=C(C(=C1)S(=O)(=O)N)Cl |
Specifications
| 6961-82-6 | |
| White-Yellow | |
| 25 g | |
| JCCBZCMSYUSCFM-UHFFFAOYSA-N | |
| 2-chlorobenzenesulfonamide | |
| 81410 | |
| ≥98.0% (GC) | |
| 2-Chlorobenzenesulfonamide |
| 190°C | |
| C6H6ClNO2S | |
| o-chlorobenzenesulfonamide, 2-chlorobenzene-1-sulfonamide, benzenesulfonamide, 2-chloro, unii-4bt2i28a0c, benzenesulfonamide, o-chloro, 2-chlorobenzenesulphonamide, 2-chloro-benzenesulfonamide, chlorobenzenesulfonamide, o-chlorobenzenesulphonamide, benzenesulfonamide, chloro | |
| C1=CC=C(C(=C1)S(=O)(=O)N)Cl | |
| 191.629 | |
| 191.63 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes