accessibility menu, dialog, popup

UserName

NH2SOOCl

CAS RN 6961-82-6

IUPAC Name: 2-chlorobenzene-1-sulfonamide
Synonyms: O-chlorobenzenesulfonamide
Molecular Weight (g/mol): 191.63
Molecular Formula: C6H6ClNO2S
InChi Key: JCCBZCMSYUSCFM-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=CC=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)