missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzenesulfonamide 98.0+%, TCI America™

Catalog No. C1990500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1990500G 500 g
C199025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1990500G Supplier TCI America Supplier No. C1990500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6961-82-6
Molecular Formula C6H6ClNO2S
Molecular Weight (g/mol) 191.629
InChI Key JCCBZCMSYUSCFM-UHFFFAOYSA-N
Synonym o-chlorobenzenesulfonamide, 2-chlorobenzene-1-sulfonamide, benzenesulfonamide, 2-chloro, unii-4bt2i28a0c, benzenesulfonamide, o-chloro, 2-chlorobenzenesulphonamide, 2-chloro-benzenesulfonamide, chlorobenzenesulfonamide, o-chlorobenzenesulphonamide, benzenesulfonamide, chloro
PubChem CID 81410
IUPAC Name 2-chlorobenzenesulfonamide
SMILES C1=CC=C(C(=C1)S(=O)(=O)N)Cl

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 500 g
Formula Weight 191.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorobenzenesulfonamide